Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
 

Quantofix Formaldehyde Test Kit, CTL Scientific Supply

Formaldehyde Test Kit, 0-10-20-40-60-100-200 mg/L
Catalog # 10217-516
Supplier:   Macherey-Nagel
 :  91328
Quantofix Formaldehyde Test Kit, CTL Scientific Supply
Catalog # 10217-516
Supplier:  Macherey-Nagel
CAS Number:  
Quantofix Formaldehyde Test Kit, CTL Scientific Supply
Quantofix Formaldehyde Test Kit, CTL Scientific Supply

Formaldehyde Test Kit, 0-10-20-40-60-100-200 mg/L

CAS Number:  
Price
$173.67
Log in pour la disponibilité
Each
Product Details & Documents

Quantofix Formaldehyde test strips and reagents for the semi-quantitative determination of formaldehyde in solutions

Quantofix Formaldehyde test kit has a gradation of scale: 0-10-20-40-60-100-200 mg/L HCHO beige to blue-violet.

Other aldehydes such as e.g. acetaldehyde or glutardialdehyde only react at higher concentrations, resulting in a discoloration which deviates from the color scale. Acetone does not react. Strong oxidizing and reducing agents cause low results.

 : Packaged as a box of 100, strips and reagent.

Spécifications

Spécifications

  • Désignation:
    Formaldehyde Test Kit
  • Range:
    0-10-20-40-60-100-200 mg/L
  • Cat. No.:
    10217-516
  • Storage:
    RT

Spécifications