$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Canada
ContactUsLinkComponent
STHT-C STHT-CSILICONE TUBING 5/8X7/8X1/8
10210-642
:
Holland Applied Technologies
STHT-C STHT-CSILICONE TUBING 5/8X7/8X1/8
10210-642
:
Holland Applied Technologies
:
STHT-C-625-4
Frequently Bought Together
VWR® Light-Duty Tissue Wipers
: VWR International
pH Reference Standard Buffers, VWR Chemicals BDH®
: BDH
Falcon® Centrifuge Tubes, Conical Bottom, Corning
: Corning
VWR® Disposable Petri Dishes
: VWR International
VWR® Nitrile Examination Gloves, Blue
: VWR International
VWR® Universal Pipette Tips
: VWR International
VWR® Microcentrifuge Tubes, PP
: VWR International