Stopper, Cap Design, Chemscience
Stopper, cap design, 14/20
Catalog # CA10204-596
Supplier:   CHEM SCIENCE INC CA
 :  CS-S0391420
Stopper, Cap Design, Chemscience
Catalog # CA10204-596
Stopper, Cap Design, Chemscience
Stopper, Cap Design, Chemscience
Stopper, cap design, 14/20
Price
$36.49
Log in pour la disponibilité
Case of 1
Product Details & Documents
These stoppers are designed for apparatus having an inner opening.
Spécifications
  • ST Joint No.:
    14/20
  • Désignation:
    Stopper, cap design