Schlenk Tubes, Modified, ChemScience
100 ml, Flask Modified Schlenk Tube, 19/22, PTFE
CA10204-448
: CS-F0909100
Choice of either 2 mm glass stopcock, 2 mm Teflon® stopcock or 0 to 4 mm high vacuum valve.
- Capacité:100 ml
- Désignation:Flask Modified Schlenk Tube
- ST Joint Size:19/22
- Stopcock Type:PTFE



