Schlenk Tubes, Modified, ChemScience
100 ml, Flask Modified Schlenk Tube, 14/20, PTFE
Catalog # CA10204-446
Supplier: CHEM SCIENCE INC CA
: CS-F0901100
Jump to:
Product Details & Documents
Choice of either 2 mm glass stopcock, 2 mm Teflon® stopcock or 0 to 4 mm high vacuum valve.
Spécifications
- Capacité:100 ml
- Désignation:Flask Modified Schlenk Tube
- ST Joint Size:14/20
- Stopcock Type:PTFE