Schlenk Tubes, Modified, ChemScience
50 ml, Flask Modified Schlenk Tube, 24/40, PTFE
CA10204-444
: CS-F0902450
Choice of either 2 mm glass stopcock, 2 mm Teflon® stopcock or 0 to 4 mm high vacuum valve.
- Capacité:50 ml
- Désignation:Flask Modified Schlenk Tube
- ST Joint Size:24/40
- Stopcock Type:PTFE