Round-Bottom Flasks with Four Vertical Necks, ChemScience | Avantor
Round-Bottom Flasks with Four Vertical Necks, ChemScience
2000 ml, Flask, Round Bottom, 4-Neck Vertical, 34/45, 24/40
Catalog # CA10204-414
Supplier:   CHEM SCIENCE INC CA
Fournisseur #:  CS-F021342L
Round-Bottom Flasks with Four Vertical Necks, ChemScience
Catalog # CA10204-414
Round-Bottom Flasks with Four Vertical Necks, ChemScience
Round-Bottom Flasks with Four Vertical Necks, ChemScience
2000 ml, Flask, Round Bottom, 4-Neck Vertical, 34/45, 24/40
Price
542,20 $
Log in pour la disponibilité
Case of 1
Product Details & Documents
With identical vertical side joints.
Spécifications
  • Capacité:
    2000 ml
  • Désignation:
    Flask, Round Bottom, 4-Neck Vertical
  • Side ST Joint Size:
    24/40
  • Center Joint:
    34/45