Round-Bottom Flasks with Four Vertical Necks, ChemScience
500 ml, Flask, Round Bottom, 4-Neck Vertical, 29/42, 24/40
  CA10204-400
 :  CS-F0212500
Round-Bottom Flasks with Four Vertical Necks, ChemScience
  CA10204-400
Round-Bottom Flasks with Four Vertical Necks, ChemScience
Round-Bottom Flasks with Four Vertical Necks, ChemScience
500 ml, Flask, Round Bottom, 4-Neck Vertical, 29/42, 24/40
$341.37
Case of 1
With identical vertical side joints.
  • Capacité:
    500 ml
  • Désignation:
    Flask, Round Bottom, 4-Neck Vertical
  • Side ST Joint Size:
    24/40
  • Center Joint:
    29/42