Round-Bottom Flasks with Four Vertical Necks, ChemScience
500 ml, Flask, Round Bottom, 4-Neck Vertical, 29/42, 24/40
CA10204-400
: CS-F0212500
With identical vertical side joints.
- Capacité:500 ml
- Désignation:Flask, Round Bottom, 4-Neck Vertical
- Side ST Joint Size:24/40
- Center Joint:29/42