Round-Bottom Flasks with Four Vertical Necks, ChemScience | Avantor
Round-Bottom Flasks with Four Vertical Necks, ChemScience
500 ml, Flask, Round Bottom, 4-Neck Vertical, 19/22, 14/20
Catalog # CA10204-394
Supplier:   CHEM SCIENCE INC CA
Fournisseur #:  CS-F0219500
Round-Bottom Flasks with Four Vertical Necks, ChemScience
Catalog # CA10204-394
Round-Bottom Flasks with Four Vertical Necks, ChemScience
Round-Bottom Flasks with Four Vertical Necks, ChemScience
500 ml, Flask, Round Bottom, 4-Neck Vertical, 19/22, 14/20
Price
$293.70
Log in pour la disponibilité
Case of 1
Product Details & Documents
With identical vertical side joints.
Spécifications
  • Capacité:
    500 ml
  • Désignation:
    Flask, Round Bottom, 4-Neck Vertical
  • Side ST Joint Size:
    14/20
  • Center Joint:
    19/22