Round-Bottom Flasks with Four Vertical Necks, ChemScience
500 ml, Flask, Round Bottom, 4-Neck Vertical, 19/22, 14/20
Catalog # CA10204-394
Supplier: CHEM SCIENCE INC CA
Fournisseur #: CS-F0219500
Round-Bottom Flasks with Four Vertical Necks, ChemScience
Catalog # CA10204-394
Supplier: CHEM SCIENCE INC CA
Jump to:
Product Details & Documents
With identical vertical side joints.
Spécifications
- Capacité:500 ml
- Désignation:Flask, Round Bottom, 4-Neck Vertical
- Side ST Joint Size:14/20
- Center Joint:19/22