Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
Schlenk Tubes, Modified, ChemScience

Schlenk Tubes, Modified, ChemScience

1000 ml, Flask Modified Schlenk Tube, 24/40, Glass
Catalog # CA10203-980
Supplier:   CHEM SCIENCE INC CA
Supplier #:  CS-F0904000G
Schlenk Tubes, Modified, ChemScience
Catalog # CA10203-980
Schlenk Tubes, Modified, ChemScience
Schlenk Tubes, Modified, ChemScience

1000 ml, Flask Modified Schlenk Tube, 24/40, Glass

Price
$232.25
Log in for availability
Case of 1
Product Details & Documents

Choice of either 2 mm glass stopcock, 2 mm Teflon® stopcock or 0 to 4 mm high vacuum valve.

Specifications
  • Capacité:
    1000 ml
  • Désignation:
    Flask Modified Schlenk Tube
  • ST Joint Size:
    24/40
  • Stopcock Type:
    Glass