Flasks, Heavy Wall, Pear Shaped, Single Neck, ChemScience
200 ml, 24/40
Catalog # CA10203-970
Supplier: CHEM SCIENCE INC CA
: CS-F0324200
Flasks, Heavy Wall, Pear Shaped, Single Neck, ChemScience
Catalog # CA10203-970
Supplier: CHEM SCIENCE INC CA
Jump to:
Product Details & Documents
Flasks feature a pear shape with single neck.
- Heavy wall design
Spécifications
- Capacité:200 ml
- ST Joint Size:24/40