Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
Vacuum Traps, Spherical Joints, Chemscience | Avantor

Vacuum Traps, Spherical Joints, Chemscience

Vacuum trap, spherical joints, 35/20
Catalog # CA10203-022
Supplier:   CHEM SCIENCE INC CA
Fournisseur #:  CS-V0175039
Vacuum Traps, Spherical Joints, Chemscience
Catalog # CA10203-022
Vacuum Traps, Spherical Joints, Chemscience
Vacuum Traps, Spherical Joints, Chemscience

Vacuum trap, spherical joints, 35/20

Price
$53.07
Log in pour la disponibilité
Case of 1
Product Details & Documents

With 35/20 spherical joints for added flexibility.

  • Trap body has 50 mm O.D. and 360 mm length

 : Order the three adapters and 35/20 SS pinch clamps separately.

Spécifications
  • ST Joint No.:
    35/20
  • Désignation:
    Vacuum trap, spherical joints
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.