Stopcocks, PTFE Straight Bore, Chemscience | Avantor
Stopcocks, PTFE Straight Bore, Chemscience
Stopcock, PTFE straight bore, 6 mm, 16/35
Catalog # CA10202-964
Supplier:   CHEM SCIENCE INC CA
Fournisseur #:  CS-S0101506
Stopcocks, PTFE Straight Bore, Chemscience
Catalog # CA10202-964
Stopcocks, PTFE Straight Bore, Chemscience
Stopcocks, PTFE Straight Bore, Chemscience
Stopcock, PTFE straight bore, 6 mm, 16/35
Price
$158.03
Log in pour la disponibilité
Case of 1
Product Details & Documents
With 1:5 PTFE plug and a threaded pressure nut.
Stopcock barrels are polished to prevent PTFE plug from scratch.
Spécifications
  • Désignation:
    Stopcock, PTFE straight bore
  • Bore Size:
    6 mm
  • Plug Size:
    16/35