Stopcocks, PTFE Straight Bore, Chemscience
Stopcock, PTFE straight bore, 6 mm, 16/35
Catalog # CA10202-964
Supplier: CHEM SCIENCE INC CA
Fournisseur #: CS-S0101506
Jump to:
Product Details & Documents
With 1:5 PTFE plug and a threaded pressure nut.
Stopcock barrels are polished to prevent PTFE plug from scratch.
Spécifications
- Désignation:Stopcock, PTFE straight bore
- Bore Size:6 mm
- Plug Size:16/35