Stopcocks, PTFE Straight Bore, Chemscience | Avantor
Stopcocks, PTFE Straight Bore, Chemscience
Stopcock, PTFE straight bore, 4 mm, 15.2/30
Catalog # CA10202-962
Supplier:   CHEM SCIENCE INC CA
Fournisseur #:  CS-S0101504
Stopcocks, PTFE Straight Bore, Chemscience
Catalog # CA10202-962
Stopcocks, PTFE Straight Bore, Chemscience
Stopcocks, PTFE Straight Bore, Chemscience
Stopcock, PTFE straight bore, 4 mm, 15.2/30
Price
$127.48
Log in pour la disponibilité
Case of 1
Product Details & Documents
With 1:5 PTFE plug and a threaded pressure nut.
Stopcock barrels are polished to prevent PTFE plug from scratch.
Spécifications
  • Désignation:
    Stopcock, PTFE straight bore
  • Bore Size:
    4 mm
  • Plug Size:
    15.2/30