Stopcocks, PTFE Straight Bore, Chemscience
Stopcock, PTFE straight bore, 4 mm, 15.2/30
Catalog # CA10202-962
Supplier: CHEM SCIENCE INC CA
Fournisseur #: CS-S0101504
Jump to:
Product Details & Documents
With 1:5 PTFE plug and a threaded pressure nut.
Stopcock barrels are polished to prevent PTFE plug from scratch.
Spécifications
- Désignation:Stopcock, PTFE straight bore
- Bore Size:4 mm
- Plug Size:15.2/30