Stopcocks, PTFE Straight Bore, Chemscience
Stopcock, PTFE straight bore, 2 mm, 11/25
Catalog # CA10202-960
Supplier: CHEM SCIENCE INC CA
Fournisseur #: CS-S0101502
Jump to:
Product Details & Documents
With 1:5 PTFE plug and a threaded pressure nut.
Stopcock barrels are polished to prevent PTFE plug from scratch.
Spécifications
- Désignation:Stopcock, PTFE straight bore
- Bore Size:2 mm
- Plug Size:11/25