Peptide Synthesis Vessels, Solid Phase, T-Bore, Vacuum, Chemscience
100 ml, Peptide synthesis vessel, solid phase, T-bore, vacuum, 25, 40 mm, Medium
Catalog # CA10202-846
Supplier: CHEM SCIENCE INC CA
Fournisseur #: CS-P0140100M
Peptide Synthesis Vessels, Solid Phase, T-Bore, Vacuum, Chemscience
Catalog # CA10202-846
Supplier: CHEM SCIENCE INC CA
Jump to:
Product Details & Documents
With a fritted disc of coarse or medium porosity to support resin.
- Top of the vessel has GL thread and a PTFE lined screw cap
- PTFE stopcock has a 2 mm bore with the arm being 8 mm O.D.
A connection of 8 mm O.D. is sealed on the inner joint for connection to an inner gas.
Spécifications
- Porosité:Medium
- Capacité:100 ml
- Désignation:Peptide synthesis vessel, solid phase, T-bore, vacuum
- Fritted Disc Size:40 mm
- GL Thread Size:25



