Peptide Synthesis Vessels, Solid Phase, T-Bore, Vacuum, Chemscience
50 ml, Peptide synthesis vessel, solid phase, T-bore, vacuum, 25, 25 mm, Coarse
Catalog # CA10202-840
Supplier: CHEM SCIENCE INC CA
: CS-P0140050C
Peptide Synthesis Vessels, Solid Phase, T-Bore, Vacuum, Chemscience
Catalog # CA10202-840
Supplier: CHEM SCIENCE INC CA
Jump to:
Product Details & Documents
With a fritted disc of coarse or medium porosity to support resin.
- Top of the vessel has GL thread and a PTFE lined screw cap
- PTFE stopcock has a 2 mm bore with the arm being 8 mm O.D.
A connection of 8 mm O.D. is sealed on the inner joint for connection to an inner gas.
Spécifications
- Porosité:Coarse
- Capacité:50 ml
- Désignation:Peptide synthesis vessel, solid phase, T-bore, vacuum
- Fritted Disc Size:25 mm
- GL Thread Size:25
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



