Peptide Synthesis Vessels, Solid Phase, T-Bore, Vacuum, Chemscience
25 ml, Peptide synthesis vessel, solid phase, T-bore, vacuum, 25, 20 mm, Coarse
Catalog # CA10202-836
Supplier: CHEM SCIENCE INC CA
Fournisseur #: CS-P0140025C
Peptide Synthesis Vessels, Solid Phase, T-Bore, Vacuum, Chemscience
Catalog # CA10202-836
Supplier: CHEM SCIENCE INC CA
Jump to:
Product Details & Documents
With a fritted disc of coarse or medium porosity to support resin.
- Top of the vessel has GL thread and a PTFE lined screw cap
- PTFE stopcock has a 2 mm bore with the arm being 8 mm O.D.
A connection of 8 mm O.D. is sealed on the inner joint for connection to an inner gas.
Spécifications
- Porosité:Coarse
- Capacité:25 ml
- Désignation:Peptide synthesis vessel, solid phase, T-bore, vacuum
- Fritted Disc Size:20 mm
- GL Thread Size:25
Recommandations
Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".
Sinon, vous recevrez des recommandations génériques.



