Peptide Synthesis Vessels, Solid Phase, Chemscience
25 ml, Peptide synthesis vessel, solid phase, 25, 20 mm, Coarse
Catalog # CA10202-800
Supplier: CHEM SCIENCE INC CA
: CS-P0120025C
Peptide Synthesis Vessels, Solid Phase, Chemscience
Catalog # CA10202-800
Supplier: CHEM SCIENCE INC CA
Jump to:
Product Details & Documents
With a fritted disc of coarse or medium porosity to support resin.
- Top of the vessel has GL thread and PTFE lined screw cap
PTFE stopcock has a 2 mm bore with the arm being 8 mm O.D.
Spécifications
- Porosité:Coarse
- Capacité:25 ml
- Désignation:Peptide synthesis vessel, solid phase
- Fritted Disc Size:20 mm
- GL Thread Size:25



