Schlenk Tubes, Modified, ChemScience
1000 ml, Flask Modified Schlenk Tube, 24/40, HV valve
CA10202-304
: CS-F0904000H
Choice of either 2 mm glass stopcock, 2 mm Teflon® stopcock or 0 to 4 mm high vacuum valve.
- Capacité:1000 ml
- Désignation:Flask Modified Schlenk Tube
- ST Joint Size:24/40
- Stopcock Type:HV valve



