Schlenk Tubes, Modified, ChemScience
200 ml, Flask Modified Schlenk Tube, 14/20, Glass
CA10202-296
: CS-F0901200G
Choice of either 2 mm glass stopcock, 2 mm Teflon® stopcock or 0 to 4 mm high vacuum valve.
- Capacité:200 ml
- Désignation:Flask Modified Schlenk Tube
- ST Joint Size:14/20
- Stopcock Type:Glass



