Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
 

Schlenk Tubes, Modified, ChemScience

100 ml, Flask Modified Schlenk Tube, 14/20, Glass
Catalog # CA10202-290
Supplier:   CHEM SCIENCE INC CA
 :  CS-F0901100G
Schlenk Tubes, Modified, ChemScience
Catalog # CA10202-290
Schlenk Tubes, Modified, ChemScience
Schlenk Tubes, Modified, ChemScience

100 ml, Flask Modified Schlenk Tube, 14/20, Glass

Price
$162.63
Log in pour la disponibilité
Case of 1
Product Details & Documents

Choice of either 2 mm glass stopcock, 2 mm Teflon® stopcock or 0 to 4 mm high vacuum valve.

Spécifications
  • Capacité:
    100 ml
  • Désignation:
    Flask Modified Schlenk Tube
  • ST Joint Size:
    14/20
  • Stopcock Type:
    Glass