Flasks, Heavy Wall, Pear Shaped, Single Neck, ChemScience
Flasks, Heavy Wall, Pear Shaped, Single Neck, ChemScience
15 ml, 14/20
Catalog # CA10202-168
Supplier:   CHEM SCIENCE INC CA
Supplier #:  CS-F0321415
Flasks, Heavy Wall, Pear Shaped, Single Neck, ChemScience
Catalog # CA10202-168
Flasks, Heavy Wall, Pear Shaped, Single Neck, ChemScience
Flasks, Heavy Wall, Pear Shaped, Single Neck, ChemScience
15 ml, 14/20
Price
$94.43
Log in pour la disponibilité
Case of 1
Product Details & Documents
Flasks feature a pear shape with single neck.
  • Heavy wall design
Spécifications
  • Capacité:
    15 ml
  • ST Joint Size:
    14/20