Flasks, Flat Bottom, Amber, Heavy Wall, ChemScience | Avantor
Flasks, Flat Bottom, Amber, Heavy Wall, ChemScience
Flat Bottom Flask, Amber, 500 ml, 24/40
Catalog # CA10202-080
Supplier:   CHEM SCIENCE INC CA
Fournisseur #:  CS-F0124500A
Flasks, Flat Bottom, Amber, Heavy Wall, ChemScience
Catalog # CA10202-080
Flasks, Flat Bottom, Amber, Heavy Wall, ChemScience
Flasks, Flat Bottom, Amber, Heavy Wall, ChemScience
Flat Bottom Flask, Amber, 500 ml, 24/40
Price
$172.39
Log in pour la disponibilité
Case of 1
Product Details & Documents
Heavy wall single neck, flat bottom flask with an outer joint.
Spécifications
  • Capacité:
    500 ml
  • Désignation:
    Flat Bottom Flask, Amber
  • Joint Size:
    24/40