Round-Bottom Flasks with Two Angled Necks, Heavy Wall, ChemScience
Round-Bottom Flasks with Two Angled Necks, Heavy Wall, ChemScience
500 ml, Flask, Two Neck, Angled, 24/40, 24/40
Catalog # CA10201-884
Supplier:   CHEM SCIENCE INC CA
Supplier #:  CS-F0414504
Round-Bottom Flasks with Two Angled Necks, Heavy Wall, ChemScience
Catalog # CA10201-884
Round-Bottom Flasks with Two Angled Necks, Heavy Wall, ChemScience
Round-Bottom Flasks with Two Angled Necks, Heavy Wall, ChemScience
500 ml, Flask, Two Neck, Angled, 24/40, 24/40
Price
$240.40
Log in for availability
Case of 1
Product Details & Documents
Round bottom flask with joints.
  • Heavy wall design
All sizes listed below are hand-blown from tubing to ensure uniform wall thickness.
Specifications
  • Capacité:
    500 ml
  • Désignation:
    Flask, Two Neck, Angled
  • Center Neck [ST] Joint:
    24/40
  • Side Neck ST Joints:
    24/40