Round-Bottom Flasks with Two Angled Necks, Heavy Wall, ChemScience
500 ml, Flask, Two Neck, Angled, 24/40, 24/40
Catalog # CA10201-884
Supplier: CHEM SCIENCE INC CA
Supplier #: CS-F0414504
Round-Bottom Flasks with Two Angled Necks, Heavy Wall, ChemScience
Catalog # CA10201-884
Supplier: CHEM SCIENCE INC CA
Jump to:
Product Details & Documents
Round bottom flask with joints.
- Heavy wall design
All sizes listed below are hand-blown from tubing to ensure uniform wall thickness.
Specifications
- Capacité:500 ml
- Désignation:Flask, Two Neck, Angled
- Center Neck [ST] Joint:24/40
- Side Neck ST Joints:24/40