Round-Bottom Flasks with Two Angled Necks, Heavy Wall, ChemScience
250 ml, Flask, Two Neck, Angled, 19/22, 14/20
Catalog # CA10201-864
Supplier: CHEM SCIENCE INC CA
: CS-F0410250
Round-Bottom Flasks with Two Angled Necks, Heavy Wall, ChemScience
Catalog # CA10201-864
Supplier: CHEM SCIENCE INC CA
Jump to:
Product Details & Documents
Round bottom flask with joints.
- Heavy wall design
All sizes listed below are hand-blown from tubing to ensure uniform wall thickness.
Specifications
- Capacité:250 ml
- Désignation:Flask, Two Neck, Angled
- Center Neck [ST] Joint:19/22
- Side Neck ST Joints:14/20