Round-Bottom Flasks with Two Angled Necks, Heavy Wall, ChemScience
250 ml, Flask, Two Neck, Angled, 19/22, 14/20
Catalog # CA10201-864
Supplier:   CHEM SCIENCE INC CA
 :  CS-F0410250
Round-Bottom Flasks with Two Angled Necks, Heavy Wall, ChemScience
Catalog # CA10201-864
Round-Bottom Flasks with Two Angled Necks, Heavy Wall, ChemScience
Round-Bottom Flasks with Two Angled Necks, Heavy Wall, ChemScience
250 ml, Flask, Two Neck, Angled, 19/22, 14/20
Price
$178.45
Log in for availability
Case of 1
Product Details & Documents
Round bottom flask with joints.
  • Heavy wall design
All sizes listed below are hand-blown from tubing to ensure uniform wall thickness.
Specifications
  • Capacité:
    250 ml
  • Désignation:
    Flask, Two Neck, Angled
  • Center Neck [ST] Joint:
    19/22
  • Side Neck ST Joints:
    14/20