Round-Bottom Flasks with Two Angled Necks, Heavy Wall, ChemScience
100 ml, Flask, Two Neck, Angled, 19/22, 14/20
  CA10201-852
 :  CS-F0411900
Round-Bottom Flasks with Two Angled Necks, Heavy Wall, ChemScience
  CA10201-852
Round-Bottom Flasks with Two Angled Necks, Heavy Wall, ChemScience
Round-Bottom Flasks with Two Angled Necks, Heavy Wall, ChemScience
100 ml, Flask, Two Neck, Angled, 19/22, 14/20
162,87 $
Case of 1
Round bottom flask with joints.
  • Heavy wall design
All sizes listed below are hand-blown from tubing to ensure uniform wall thickness.
  • Capacité:
    100 ml
  • Désignation:
    Flask, Two Neck, Angled
  • Center Neck [ST] Joint:
    19/22
  • Side Neck ST Joints:
    14/20