Round-Bottom Flasks with Two Angled Necks, Heavy Wall, ChemScience
100 ml, Flask, Two Neck, Angled, 19/22, 14/20
CA10201-852
: CS-F0411900
Round-Bottom Flasks with Two Angled Necks, Heavy Wall, ChemScience
CA10201-852
Round bottom flask with joints.
- Heavy wall design
All sizes listed below are hand-blown from tubing to ensure uniform wall thickness.
- Capacité:100 ml
- Désignation:Flask, Two Neck, Angled
- Center Neck [ST] Joint:19/22
- Side Neck ST Joints:14/20