Round-Bottom Flasks with Two Angled Necks, Heavy Wall, ChemScience
50 ml, Flask, Two Neck, Angled, 24/40, 19/22
CA10201-846
: CS-F0414950
Round-Bottom Flasks with Two Angled Necks, Heavy Wall, ChemScience
CA10201-846
Round bottom flask with joints.
- Heavy wall design
All sizes listed below are hand-blown from tubing to ensure uniform wall thickness.
- Capacité:50 ml
- Désignation:Flask, Two Neck, Angled
- Center Neck [ST] Joint:24/40
- Side Neck ST Joints:19/22