Round-Bottom Flasks with Two Angled Necks, Heavy Wall, ChemScience
50 ml, Flask, Two Neck, Angled, 24/40, 19/22
  CA10201-846
 :  CS-F0414950
Round-Bottom Flasks with Two Angled Necks, Heavy Wall, ChemScience
  CA10201-846
Round-Bottom Flasks with Two Angled Necks, Heavy Wall, ChemScience
Round-Bottom Flasks with Two Angled Necks, Heavy Wall, ChemScience
50 ml, Flask, Two Neck, Angled, 24/40, 19/22
$252.21
Case of 1
Round bottom flask with joints.
  • Heavy wall design
All sizes listed below are hand-blown from tubing to ensure uniform wall thickness.
  • Capacité:
    50 ml
  • Désignation:
    Flask, Two Neck, Angled
  • Center Neck [ST] Joint:
    24/40
  • Side Neck ST Joints:
    19/22