Flasks, Heavy Wall, Round Bottom, Single Neck With Hooks, ChemScience
25 ml, Round Bottom Flask with Hooks, 19/22
Catalog # CA10201-840
Supplier: CHEM SCIENCE INC CA
Fournisseur #: CS-F0331925
Flasks, Heavy Wall, Round Bottom, Single Neck With Hooks, ChemScience
Catalog # CA10201-840
Supplier: CHEM SCIENCE INC CA
Jump to:
Product Details & Documents
Heavy wall design.
Spécifications
- Capacité:25 ml
- Désignation:Round Bottom Flask with Hooks
- Joint Size:19/22