Flasks, Heavy Wall, Round Bottom, Threaded Inlet, ChemScience
500 ml, Round Bottom Flask, Threaded Inlet, 24/40
Catalog # CA10201-828
Supplier:   CHEM SCIENCE INC CA
 :  CS-F0144500
Flasks, Heavy Wall, Round Bottom, Threaded Inlet, ChemScience
Catalog # CA10201-828
Flasks, Heavy Wall, Round Bottom, Threaded Inlet, ChemScience
Flasks, Heavy Wall, Round Bottom, Threaded Inlet, ChemScience
500 ml, Round Bottom Flask, Threaded Inlet, 24/40
Price
$151.28
Log in pour la disponibilité
Case of 1
Product Details & Documents
All sizes listed below are hand-blown from tubing to ensure uniform wall thickness.
  • An outer diameter between 5 and 7.5 mm
  • Heavy wall design
With a threaded side port for insertion of thermometer or bleed tubes.
Spécifications
  • Capacité:
    500 ml
  • Désignation:
    Round Bottom Flask, Threaded Inlet
  • Joint Size:
    24/40