Flasks, Heavy Wall, Round Bottom, Threaded Inlet, ChemScience
250 ml, Round Bottom Flask, Threaded Inlet, 14/20
Catalog # CA10201-822
Supplier: CHEM SCIENCE INC CA
Supplier #: CS-F0141250
Flasks, Heavy Wall, Round Bottom, Threaded Inlet, ChemScience
Catalog # CA10201-822
Supplier: CHEM SCIENCE INC CA
Jump to:
Product Details & Documents
All sizes listed below are hand-blown from tubing to ensure uniform wall thickness.
- An outer diameter between 5 and 7.5 mm
- Heavy wall design
With a threaded side port for insertion of thermometer or bleed tubes.
Specifications
- Capacité:250 ml
- Désignation:Round Bottom Flask, Threaded Inlet
- Joint Size:14/20