Flasks, Heavy Wall, Round Bottom, Threaded Inlet, ChemScience
Flasks, Heavy Wall, Round Bottom, Threaded Inlet, ChemScience
250 ml, Round Bottom Flask, Threaded Inlet, 14/20
Catalog # CA10201-822
Supplier:   CHEM SCIENCE INC CA
Supplier #:  CS-F0141250
Flasks, Heavy Wall, Round Bottom, Threaded Inlet, ChemScience
Catalog # CA10201-822
Flasks, Heavy Wall, Round Bottom, Threaded Inlet, ChemScience
Flasks, Heavy Wall, Round Bottom, Threaded Inlet, ChemScience
250 ml, Round Bottom Flask, Threaded Inlet, 14/20
Price
$127.83
Log in for availability
Case of 1
Product Details & Documents
All sizes listed below are hand-blown from tubing to ensure uniform wall thickness.
  • An outer diameter between 5 and 7.5 mm
  • Heavy wall design
With a threaded side port for insertion of thermometer or bleed tubes.
Specifications
  • Capacité:
    250 ml
  • Désignation:
    Round Bottom Flask, Threaded Inlet
  • Joint Size:
    14/20
Recommandations
Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".
Sinon, vous recevrez des recommandations génériques.