Flasks, Heavy Wall, Round Bottom, Threaded Inlet, ChemScience
100 ml, Round Bottom Flask, Threaded Inlet, 24/40
Catalog # CA10201-820
Supplier: CHEM SCIENCE INC CA
Fournisseur #: CS-F0144100
Flasks, Heavy Wall, Round Bottom, Threaded Inlet, ChemScience
Catalog # CA10201-820
Supplier: CHEM SCIENCE INC CA
Jump to:
Product Details & Documents
All sizes listed below are hand-blown from tubing to ensure uniform wall thickness.
- An outer diameter between 5 and 7.5 mm
- Heavy wall design
With a threaded side port for insertion of thermometer or bleed tubes.
Spécifications
- Capacité:100 ml
- Désignation:Round Bottom Flask, Threaded Inlet
- Joint Size:24/40