Flasks, Heavy Wall, Round Bottom, Threaded Inlet, ChemScience | Avantor
Flasks, Heavy Wall, Round Bottom, Threaded Inlet, ChemScience
100 ml, Round Bottom Flask, Threaded Inlet, 19/22
Catalog # CA10201-818
Supplier:   CHEM SCIENCE INC CA
Fournisseur #:  CS-F0149100
Flasks, Heavy Wall, Round Bottom, Threaded Inlet, ChemScience
Catalog # CA10201-818
Flasks, Heavy Wall, Round Bottom, Threaded Inlet, ChemScience
Flasks, Heavy Wall, Round Bottom, Threaded Inlet, ChemScience
100 ml, Round Bottom Flask, Threaded Inlet, 19/22
Price
$116.43
Log in pour la disponibilité
Case of 1
Product Details & Documents
All sizes listed below are hand-blown from tubing to ensure uniform wall thickness.
  • An outer diameter between 5 and 7.5 mm
  • Heavy wall design
With a threaded side port for insertion of thermometer or bleed tubes.
Spécifications
  • Capacité:
    100 ml
  • Désignation:
    Round Bottom Flask, Threaded Inlet
  • Joint Size:
    19/22