Flasks, Heavy Wall, Round Bottom, Threaded Inlet, ChemScience
100 ml, Round Bottom Flask, Threaded Inlet, 14/20
CA10201-816
: CS-F0141100
All sizes listed below are hand-blown from tubing to ensure uniform wall thickness.
- An outer diameter between 5 and 7.5 mm
- Heavy wall design
With a threaded side port for insertion of thermometer or bleed tubes.
- Capacité:100 ml
- Désignation:Round Bottom Flask, Threaded Inlet
- Joint Size:14/20