Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
Flasks, Heavy Wall, Round Bottom, Threaded Inlet, ChemScience | Avantor

Flasks, Heavy Wall, Round Bottom, Threaded Inlet, ChemScience

50 ml, Round Bottom Flask, Threaded Inlet, 19/22
Catalog # CA10201-814
Supplier:   CHEM SCIENCE INC CA
Fournisseur #:  CS-F0141950
Flasks, Heavy Wall, Round Bottom, Threaded Inlet, ChemScience
Catalog # CA10201-814
Flasks, Heavy Wall, Round Bottom, Threaded Inlet, ChemScience
Flasks, Heavy Wall, Round Bottom, Threaded Inlet, ChemScience

50 ml, Round Bottom Flask, Threaded Inlet, 19/22

Price
100,73 $
Log in pour la disponibilité
Case of 1
Product Details & Documents

All sizes listed below are hand-blown from tubing to ensure uniform wall thickness.

  • An outer diameter between 5 and 7.5 mm
  • Heavy wall design

With a threaded side port for insertion of thermometer or bleed tubes.

Spécifications
  • Capacité:
    50 ml
  • Désignation:
    Round Bottom Flask, Threaded Inlet
  • Joint Size:
    19/22
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.