Flasks, Heavy Wall, Round Bottom, Single Neck, Spherical Joint, ChemScience
500 ml, Spherical Joint Round Bottom Flask, 35/20
Catalog # CA10201-804
Supplier: CHEM SCIENCE INC CA
Fournisseur #: CS-F0527500
Flasks, Heavy Wall, Round Bottom, Single Neck, Spherical Joint, ChemScience
Catalog # CA10201-804
Supplier: CHEM SCIENCE INC CA
Jump to:
Product Details & Documents
Heavy wall design.
All sizes listed below are hand-blown from tubing to ensure uniform wall thickness.
Spécifications
- Capacité:500 ml
- Désignation:Spherical Joint Round Bottom Flask
- Joint Size:35/20