Flasks, Heavy Wall, Round Bottom, Single Neck, Spherical Joint, ChemScience | Avantor
Flasks, Heavy Wall, Round Bottom, Single Neck, Spherical Joint, ChemScience
500 ml, Spherical Joint Round Bottom Flask, 35/20
Catalog # CA10201-804
Supplier:   CHEM SCIENCE INC CA
Fournisseur #:  CS-F0527500
Flasks, Heavy Wall, Round Bottom, Single Neck, Spherical Joint, ChemScience
Catalog # CA10201-804
Flasks, Heavy Wall, Round Bottom, Single Neck, Spherical Joint, ChemScience
Flasks, Heavy Wall, Round Bottom, Single Neck, Spherical Joint, ChemScience
500 ml, Spherical Joint Round Bottom Flask, 35/20
Price
71,15 $
Log in pour la disponibilité
Case of 1
Product Details & Documents
Heavy wall design.
All sizes listed below are hand-blown from tubing to ensure uniform wall thickness.
Spécifications
  • Capacité:
    500 ml
  • Désignation:
    Spherical Joint Round Bottom Flask
  • Joint Size:
    35/20