Flasks, Heavy Wall, Round Bottom, Single Neck, Spherical Joint, ChemScience
250 ml, Spherical Joint Round Bottom Flask, 35/20
CA10201-802
: CS-F0527250
Flasks, Heavy Wall, Round Bottom, Single Neck, Spherical Joint, ChemScience
CA10201-802
Heavy wall design.
All sizes listed below are hand-blown from tubing to ensure uniform wall thickness.
- Capacité:250 ml
- Désignation:Spherical Joint Round Bottom Flask
- Joint Size:35/20