Quick Release Iris Hook, OR Grade, Sklar
Quick Release Iris Hook, OR Grade, Sklar
30.5 cm (12"), Stainless steel, Quick Release
Catalog # 10199-170
Supplier:   Sklar
Supplier #:  90-8348
Quick Release Iris Hook, OR Grade, Sklar
Catalog # 10199-170
Supplier:  Sklar
Quick Release Iris Hook, OR Grade, Sklar
Quick Release Iris Hook, OR Grade, Sklar
30.5 cm (12"), Stainless steel, Quick Release
Price
$428.72
New Customer Promotional Price
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
The Sklar Iris Hook is a versatile instrument that can be used in many procedures to hold and retract tissue.
It is similar to, but smaller than the Emmett Hook, and the hook is slightly angled. The hook, available in multiple diameters, is at the end of a long, narrow handle, often 9 or 12 inches in length.
Specifications
  • Longueur:
    30.5 cm (12")
  • Matière:
    Stainless steel
  • Tip Configuration:
    Quick Release