Quick Release Iris Hook, OR Grade, Sklar
The Sklar Iris Hook is a versatile instrument that can be used in many procedures to hold and retract tissue.
It is similar to, but smaller than the Emmett Hook, and the hook is slightly angled. The hook, available in multiple diameters, is at the end of a long, narrow handle, often 9 or 12 inches in length.
- Longueur:22.9 cm (9")
- Matière:Stainless steel
- Tip Configuration:Quick Release