pH Buffer Solutions, Hach
±0.02, 25 °C, 15 mL, 10.01, Blue
Catalog # 10199-510
Supplier #: 2283636
Jump to:
Discontinued
Le produit n'est plus fabriqué. Cependant, nous avons trouvé des produits alternatifs similaires.
Alternatives
BDH BUFFER PH 10 BLUE 20X20ML Catalog #BDH5070-20ML
Product BDH5070-20ML is similar to 10199-510 but may have a different manufacturer and/or packaging.
BDH BUFFER REF STD PH10 BLUE 500ML Catalog #BDH5072-500ML
Product BDH5072-500ML is similar to 10199-510 but may have a different manufacturer and/or packaging.
PH 10.01 BUFFER COLOR-BLUE 5X60ML Catalog #23198-224
Product 23198-224 is similar to 10199-510 but may have a different manufacturer and/or packaging.
Product Details & Documents
For pH meter standardization
Available colorless, or color-coded for easy identification.
Documents
Certificates
Recherche par numéro de lotSpécifications
Spécifications
- Color:Blue
- Taille:15 mL
- Reference Temperature:25 °C
- pH:10.01
- pH Accuracy:±0.02



