Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
pH Buffer Solutions, Hach

pH Buffer Solutions, Hach

±0.02, 25 °C, 15 mL, 7.00, Yellow
Catalog # 10199-506
Supplier #:  2283536
pH Buffer Solutions, Hach
Catalog # 10199-506
Supplier:  
CAS Number:  
pH Buffer Solutions, Hach
pH Buffer Solutions, Hach

±0.02, 25 °C, 15 mL, 7.00, Yellow

CAS Number:  
Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
BUFFER SOLN YELLOW PH7.00 15ML Catalog #CA10199-506

Product CA10199-506 is a replacement for 10199-506 and has the same packaging and specifications, but may be a different quantity.

Product Details & Documents

For pH meter standardization

Available colorless, or color-coded for easy identification.

Documents

Specifications

Specifications

  • Color:
    Yellow
  • Taille:
    15 mL
  • Reference Temperature:
    25 °C
  • pH:
    7.00
  • pH Accuracy:
    ±0.02
  • Cat. No.:
    10199-506

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.