Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
 

pH Buffer Solutions, Hach

±0.02, 25 °C, 50 mL, 7.00, Yellow
Catalog # 10199-504
pH Buffer Solutions, Hach
Catalog # 10199-504
Supplier:  
CAS Number:  
pH Buffer Solutions, Hach
pH Buffer Solutions, Hach

±0.02, 25 °C, 50 mL, 7.00, Yellow

CAS Number:  
Discontinued
Le produit n'est plus fabriqué. Cependant, nous avons trouvé des produits alternatifs similaires.
Alternatives
BUFFER SOLN YELLOW PH 7.00 50ML Catalog #CA10199-504

 

Product Details & Documents

For pH meter standardization

Available colorless, or color-coded for easy identification.

Spécifications

Spécifications

  • Color:
    Yellow
  • Taille:
    50 mL
  • Reference Temperature:
    25 °C
  • pH:
    7.00
  • pH Accuracy:
    ±0.02

Spécifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.