Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
pH Buffer Solutions, Hach | Avantor

pH Buffer Solutions, Hach

±0.02, 25 °C, 15 mL, 4.01, Colorless
Catalog # 10199-498
pH Buffer Solutions, Hach
Catalog # 10199-498
Supplier:  
CAS Number:  
pH Buffer Solutions, Hach
pH Buffer Solutions, Hach

±0.02, 25 °C, 15 mL, 4.01, Colorless

CAS Number:  
Discontinued
Product Details & Documents

For pH meter standardization

Available colorless, or color-coded for easy identification.

Documents

Spécifications

Spécifications

  • Color:
    Colorless
  • Taille:
    15 mL
  • Reference Temperature:
    25 °C
  • pH:
    4.01
  • pH Accuracy:
    ±0.02

Spécifications