Corning® Spheroid Microplates, Corning
Spheroid Microplates, 384, 20 - 90 µl, Round bottom, Black/Clear, Sterile, Ultra-low attachment
Supplier: Corning
Fournisseur #: 4516
Corning® Spheroid Microplates, Corning
Catalog # 10185-096
Supplier: Corning
Jump to:
Product Details & Documents
PS, black opaque microplate with optically clear bottom, with lid. The Ultra-Low Attachment surface is a covalently bound hydrogel layer that is hydrophilic and neutrally charged. The surface minimizes cell attachment, protein absorption, and enzyme activation. It is ideal for culturing stem cells, as it promotes embryoid body formation from ES cells.
- Well geometry aids formation of spheroids in centre of well
- Non cytotoxic, biologically inert and non degradable surface
- Sterilized by gamma radiation
Spécifications
- Nb de puits:384
- Sterility:Sterile
- Color:Black/Clear
- Désignation:Spheroid Microplates
- Working Volume:20 - 90 µl
- Surface Treatment:Ultra-low attachment
- Bottom Style:Round bottom
- Cat. No.:10185-096
- Environmental certification:RECs, ISO 14001 - Kennebunk
- Environmentally Preferable:Low Manufacturing Impact
Recommandations
Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".
Sinon, vous recevrez des recommandations génériques.



