Staticmaster® EZ Mounts, NRD
Staticmaster 1" EZ mount for use with 1U400 ionizing cartridge
Catalog # 10183-840
Supplier: NRD Staticmaster
Fournisseur #: BF4
Jump to:
Product Details & Documents
An indispensable accessory for mounting a 2U500 or 1U400 Ionizing Cartridge in the balance chamber.
- Simple to use
- Makes installation of ionizing cartridges fast and easy
Simply remove the protective layer of the double-sided tape and affix the E-Z mount to the back wall of the chamber. Once the mount is attached, installing ionizing cartridges is fast and easy.
Spécifications
- Désignation:Staticmaster 1" EZ mount for use with 1U400 ionizing cartridge
Recommandations
Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".
Sinon, vous recevrez des recommandations génériques.