Dendritic Cell Medium, PromoCell
DC Generation Medium DXF (Ready-to-use with Cytokines), 250 ml
Catalog # 10172-400
Supplier:   Promocell
 :  C-28052
Dendritic Cell Medium, PromoCell
Catalog # 10172-400
Supplier:  Promocell
CAS Number:  
Dendritic Cell Medium, PromoCell
Dendritic Cell Medium, PromoCell
DC Generation Medium DXF (Ready-to-use with Cytokines), 250 ml
CAS Number:  
Price
2 656,03 $
Log in pour la disponibilité
Each (250ml)
  • L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.
  • Return Policy
Product Details & Documents
The PromoCell Dendritic Cell (DC) Generation Medium has been developed for the easy and efficient generation of immature as well as fully mature myeloid Dendritic Cells from peripheral blood monocytes
The DC Generation Medium DXF is a chemically defined and xeno-free formulation for use with freshly isolated cells. It is free of all animal derived components and substances of human origin with Human Serum Albumin as the only exception. The DC Base Media do not include cytokines and must therefore be supplemented according to the user’s individual needs. The Monocyte Attachment Medium allows for the efficient adherence selection of Monocytes from freshly isolated Mononuclear Cells (MNC) while maintaining optimal cell health. Thus, the time-consuming and costly immunomagnetic purification of monocytes prior to dendritic cell generation is not necessary and can be skipped.
Recommended for Human Monocytes, freshly isolated and cryopreserved, Human Mononuclear Cells, freshly isolated
PromoCell DC Generation Medium allows for the generation of DC from cryopreserved as well as freshly isolated cells. In the latter case, the Monocyte Attachment Medium is also needed.
 : PromoCell DC Generation Media contain all growth factors and supplements. The DC Base Media come without cytokines and must therefore be adequately supplemented by the user. The Monocyte Attachment Medium does not need any further supplementation.
Spécifications
Spécifications
  • Désignation:
    DC Generation Medium DXF (Ready-to-use with Cytokines)
  • Taille:
    250 ml
Spécifications