Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
Absorbent Liners, Jaece

Absorbent Liners, Jaece

50.8 cm x 15 m (20" x 50') One Roll in Dispenser Box
Catalog # 10159-110
Supplier:   Jaece
Supplier #:  AL20501
Absorbent Liners, Jaece
Catalog # 10159-110
Supplier:  Jaece
Absorbent Liners, Jaece
Absorbent Liners, Jaece

50.8 cm x 15 m (20" x 50') One Roll in Dispenser Box

Price
$132.20
Log in for availability
Case of 1
Product Details & Documents

Made of highly absorbent three-ply tissue with a waterproof polypropylene backing that is resistant to most chemicals.

  • Cushions surfaces while absorbing liquids
  • Can be cut to size

Keeps spills from spreading and protects benchtops, floors, and carts.

Ordering information: Liner 82002-606 is packaged one roll per case.

Specifications
  • Désignation:
    50.8 cm x 15 m (20" x 50') One Roll in Dispenser Box
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.