EasyCurve™ Mop System, Contec®
The EasyCurve™ Mop was designed specifically for use in critical and controlled environments. It is now available with a ribbed microfiber face, ensuring smooth and easy mopping with improved particle pick-up and contamination removal.
- All components are autoclavable; ideal for a wide range of cleaning applications in the most critical sterile environments
- Lightweight mop head and handle significantly reduces operator fatigue
- Single-use mop is economical and easy to install without tools
- Curved mop and pivoting mop frame provide effortless contact with surface
- Ribbed microfiber face provides better contaminant removal
The combination of the curved mop face and the pivoting connector results in effortless surface contact and outstanding maneuverability. This makes the EasyCurve™ an ideal cleaning tool for ceilings, walls, and floors. All components of the EasyCurve™ Mop are autoclavable. Available in both nonsterile and validated sterile versions.
- Désignation:EasyCurve™ Ribbed Microfiber Mop Head
- Taille:36×11×2 cm (14×1.25×0.75")
Accessories
- DescriptionLength
- DescriptionLength
- DescriptionLength
- Description
- DescriptionLength