EasyCurve™ Mop System, Contec®
EasyCurve™ Mop System, Contec®
EasyCurve™ Ribbed Microfiber Mop Head, 36×11×2 cm (14×1.25×0.75")
  CA10153-828
 :   Contec
Supplier #:  EZMP0300
EasyCurve™ Mop System, Contec®
  CA10153-828
 :  Contec
EasyCurve™ Mop System, Contec®
EasyCurve™ Mop System, Contec®
EasyCurve™ Ribbed Microfiber Mop Head, 36×11×2 cm (14×1.25×0.75")
$1,300.00
Case of 48
The EasyCurve™ Mop was designed specifically for use in critical and controlled environments. It is now available with a ribbed microfiber face, ensuring smooth and easy mopping with improved particle pick-up and contamination removal.
  • All components are autoclavable; ideal for a wide range of cleaning applications in the most critical sterile environments
  • Lightweight mop head and handle significantly reduces operator fatigue
  • Single-use mop is economical and easy to install without tools
  • Curved mop and pivoting mop frame provide effortless contact with surface
  • Ribbed microfiber face provides better contaminant removal
The combination of the curved mop face and the pivoting connector results in effortless surface contact and outstanding maneuverability. This makes the EasyCurve™ an ideal cleaning tool for ceilings, walls, and floors. All components of the EasyCurve™ Mop are autoclavable. Available in both nonsterile and validated sterile versions.
  • Désignation:
    EasyCurve™ Ribbed Microfiber Mop Head
  • Taille:
    36×11×2 cm (14×1.25×0.75")
Accessories
  •  
    Description
    Length
  • 33502-715
    QuickConnect™ anodized aluminum telescopic handle
    127 - 234 cm (50 - 92")
    Each - $265.74
  •  
    Description
    Length
  • 33502-716
    QuickConnect™ stainless steel telescopic handle
    41 - 76 cm (16 - 30")
    Each - $502.24
  •  
    Description
    Length
  • 33502-714
    QuickConnect™ anodized aluminum telescopic handle
    41 - 76 cm (16 - 30")
    Each - $235.64
  •  
    Description
  • 89073-986
    Stainless Steel Connecting Kit for Multi-Bucket System
    Each - $127.28
  •  
    Description
    Length
  • 33502-717
    QuickConnect™ stainless steel telescopic handle
    127 - 234 cm (50 - 92")
    Each - $761.10