Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
StatSpin® Express 3 Centrifuge, Iris Sample Processing | Avantor

StatSpin® Express 3 Centrifuge, Iris Sample Processing

Centrifuge
Catalog # CA87003-180
Supplier:   Statspin
Fournisseur #:  SSX3
StatSpin® Express 3 Centrifuge, Iris Sample Processing
Catalog # CA87003-180
Supplier:  Statspin
StatSpin® Express 3 Centrifuge, Iris Sample Processing
StatSpin® Express 3 Centrifuge, Iris Sample Processing

Centrifuge

Price
$10,459.24
Log in pour la disponibilité
Each
Product Details & Documents

When rapid serum or plasma is needed, such as in coagulation testing, sample prep for stat chemistry, and cardiac profiles in the ER, this centrifuge is ideal.

  • Settings for 2, 3, and 5 minutes
  • Tubes spun in 2-minute setting equal tubes spun in 7 to 10 minute setting in other centrifuges
  • Up to 8 standard glass or plastic blood-collection tubes accepted
  • Accepted tube size: Between 1.5 to 10 ml
  • Maintenance-free, brushless motor

It has settings for 2, 3, and 5 minutes.

The centrifuge accepts up to eight standard glass or plastic blood-collection tubes from 1.5 to 10 ml. The rugged, brushless motor is maintenance free.

The two-minute setting produces serum or plasma equal to tubes spun for 7 to 10 minutes in conventional clinical centrifuges.

Certifications: UL listed. CE marked.

Ordering information: Includes 8-place rotor, removable bowl liner, universal power supply, 120 V adapter and power cord, and eight adapters each for 5 and 10 ml tubes.

Caution: Note: StatSpin centrifuges have a 6 month warranty.

Services: VWRCATALYST™ offers validation services including IQ, OQ, and PQ protocol development and execution, as well as asset management and calibration services (US only); call 888-793-2300 for more details.

Spécifications
  • Désignation:
    Centrifuge
  • Cat. No.:
    CA87003-180
  • Max. speed:
    7200 rpm
  • Acceleration/braking time:
    20 s (to 95% of speed) / <30 second s
  • Dimensions:
    18H x 26.7W x 25.4D cm (71/8 x 101/2 x 10")
  • Electrical:
    120–240V, 50/60Hz, 3A
  • Max. RCF:
    4440 g ±5%
  • Cycle time:
    2, 3, 5 min
  • Weight:
    4.3 kg (9.5 lbs.)
Accessories

Informations sur le produit

  • Catalog #
    Availability
    Price
  • Inserts for BD™ Microtubes
    Inserts for BD™ Microtubes
    CA87003-962
    Log in pour la disponibilité
    Pack of 4 - $327.05

Informations sur le produit

  • Catalog #
    Availability
    Price
  • Inserts for 7 mL (13 x 100 mm) Tubes
    Inserts for 7 mL (13 x 100 mm) Tubes
    CA87003-970
    Log in pour la disponibilité
    Pack of 8 - $313.10

Informations sur le produit

  • Catalog #
    Availability
    Price
  • Inserts for 7 mL (16 x 75 mm) Tubes
    Inserts for 7 mL (16 x 75 mm) Tubes
    CA87003-964
    Log in pour la disponibilité
    Pack of 8 - $234.70

Informations sur le produit

  • Catalog #
    Availability
    Price
  • Inserts for 5 mL (13 x 75 mm) Tubes
    Inserts for 5 mL (13 x 75 mm) Tubes
    CA87003-968
    Log in pour la disponibilité
    Pack of 8 - $453.95

Informations sur le produit

  • Catalog #
    Availability
    Price
  • Disposable Bowl Liners
    Disposable Bowl Liners
    CA87003-972
    Log in pour la disponibilité
    Pack of 3 - $265.76

Informations sur le produit

  • Catalog #
    Availability
    Price
  • Inserts for 10 mL (16 x 100 mm) Tubes
    Inserts for 10 mL (16 x 100 mm) Tubes
    CA87003-966
    Log in pour la disponibilité
    Pack of 8 - $234.70

Informations sur le produit

  • Catalog #
    Availability
    Price
  • Rotor for 1.5–10mL Tubes, 8-Place
    Rotor for 1.5–10mL Tubes, 8-Place
    CA87003-974
    Log in pour la disponibilité
    Each - $2,137.85

Recommandations

Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".

Sinon, vous recevrez des recommandations génériques.