Bucket, Rectangular, Vileda Professional-FHP
Lid for 6 L Bucket, autoclavable
Catalog # 10153-182
Supplier: Vileda Professional - FHP
Supplier #: 125948
Bucket, Rectangular, Vileda Professional-FHP
Catalog # 10153-182
Supplier: Vileda Professional - FHP
Jump to:
Product Details & Documents
High quality polypropylene buckets with 6 L capacity. Can be used individually and also on the CE Bucket Trolley or CE Pre-Prepared Trolley.
- Color coding clips included to allow differentiation in colors of buckets (red, blue, green, yellow)
- Autoclavable
Ordering information: Lid sold separately.
Specifications
- Désignation:Lid for 6 L Bucket, autoclavable