Bucket, Rectangular, Vileda Professional-FHP
Bucket, Rectangular, Vileda Professional-FHP
Lid for 6 L Bucket, autoclavable
Catalog # 10153-182
Supplier #:  125948
Bucket, Rectangular, Vileda Professional-FHP
Catalog # 10153-182
Bucket, Rectangular, Vileda Professional-FHP
Bucket, Rectangular, Vileda Professional-FHP
Lid for 6 L Bucket, autoclavable
Price
$23.30
Log in for availability
Each Case of 12
Product Details & Documents
High quality polypropylene buckets with 6 L capacity. Can be used individually and also on the CE Bucket Trolley or CE Pre-Prepared Trolley.
  • Color coding clips included to allow differentiation in colors of buckets (red, blue, green, yellow)
  • Autoclavable
Ordering information: Lid sold separately.
Specifications
  • Désignation:
    Lid for 6 L Bucket, autoclavable